missing translation for 'onlineSavingsMsg'
Learn More

Risdiplam, MedChemExpress

Product Code. 16496997
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16496997

Brand: Medchem Express HY109101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Risdiplam (RG7916) is an orally administered, centrally and peripherally distributed SMN2 pre-mRNA splicing modifier that increases survival motor neuron (SMN) protein levels.

Risdiplam (RG7916) is an orally administered, centrally and peripherally distributed SMN2 pre-mRNA splicing modifier that increases survival motor neuron (SMN) protein levels.

Specifications

Chemical Name or Material Risdiplam
Physical Form Solid
CAS 1825352-65-5
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H23N7O
For Use With (Application) Neuroscience-Neuromodulation
Synonym RG7916 RO7034067
Solubility Information Ethanol : 2 mg/mL (4.98 mM; Need ultrasonic) ∣DMSO : 2 mg/mL (4.98 mM; Need ultrasonic)
SMILES O=C(C=C(C(C=C1C)=NN2C1=NC(C)=C2)N=C3C=C4)N3C=C4N(CCN5)CC65CC6
Molecular Weight (g/mol) 401.46
Formula Weight 401.46
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.