missing translation for 'onlineSavingsMsg'
Learn More

RIPX Antibody, Novus Biologicals™

Product Code. 18328858 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328858 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328858 Supplier Novus Biologicals Supplier No. H00022902B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

RIPX Polyclonal antibody specifically detects RIPX in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPX
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_055776.1
Gene Alias KIAA0871SINGAR1, protein RUFY3, rap2 interacting protein x, Rap2-interacting protein x, RIPx, RUN and FYVE domain containing 3, singar, Singar1, Single axon-regulated protein, single axon-related 1
Host Species Mouse
Immunogen RUFY3 (NP_055776.1, 1 a.a. - 469 a.a.) full-length human protein. MSALTPPTDMPTPTTDKITQAAMETIYLCKFRVSMDGEWLCLRELDDISLTPDPEPTHEDPNYLMANERMNLMNMAKLSIKGLIESALNLGRTLDSDYAPLQQFFVVMEHCLKHGLKAKKTFLGQNKSFWGPLELVEKLVPEAAEITASVKDLPGLKTPVGRGRAWLRLALMQKKLSEYMKALINKKELLSEFYEPNALMMEEEGAIIAGLLVGLNVIDANFCMKGEDLDSQVGVIDFSMYLKDGNSSKGTEGDGQITAILDQKNYVEELNRHLNATVNNLQAKVDALEKSNTKLTEELAVANNRIITLQEEMERVKEESSYILESNRKGPKQDRTAEGQALSEARKHLKEETQLRLDVEKELEMQISMRQEMELAMKMLEKDVCEKQDALVSLRQQLDDLRALKHELAFKLQSSDLGVKQKSELNSRLEEKTNQMAATIKQLEQSEKDLVKQAKTLNSAANKLIPKHH
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22902
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.