missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£219.00 - £428.00
Specifica
| Antigen | RIPX |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|
18445681
|
Novus Biologicals
NBP1-89614-25ul |
25 μL |
£219.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18031964
|
Novus Biologicals
NBP1-89614 |
0.1 mL |
£428.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
RIPX Polyclonal specifically detects RIPX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifica
| RIPX | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 22902 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MLEKDVCEKQDALVSLRQQLDDLRALKHELAFKLQSSDLGVKQKSELNSRLEEKTNQMAATIKQLEQRLRQAERSRQSAELDNRLFKQDFGDKINSLQLEVEELTRQRNQLEL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| KIAA0871SINGAR1, protein RUFY3, rap2 interacting protein x, Rap2-interacting protein x, RIPx, RUN and FYVE domain containing 3, singar, Singar1, Single axon-regulated protein, single axon-related 1 | |
| RUFY3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto