missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£222.00
Specifications
| Antigen | RIPK5 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Beschreibung
RIPK5 Polyclonal specifically detects RIPK5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spezifikation
| RIPK5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| dual serine/threonine and tyrosine protein kinase, Dusty PK, Dusty protein kinase, DUSTYPK, KIAA0472RIP-homologous kinase, receptor interacting protein 5, receptor interacting protein kinase 5, Receptor-interacting serine/threonine-protein kinase 5, RIP5EC 2.7.12.1, RIPK5, sgK496, Sugen kinase 496 | |
| DSTYK | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04 - 0.4 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 25778 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KENELYESLMNIANRKQEEMKDMIVETLNTMKEELLDDATNMEFKDVIVPENGEPVGTREIKCCIRQIQELIISRLNQAVANKLISSVDYLRESFVGTLERCLQSLEKSQDVSVHITSNYLKQILNAAYHVEVTFHSGSS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts