missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RIPK5 Monoclonal antibody specifically detects RIPK5 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | RIPK5 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 4D5 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_056190 |
| Gene Alias | dual serine/threonine and tyrosine protein kinase, Dusty PK, Dusty protein kinase, DUSTYPK, KIAA0472RIP-homologous kinase, receptor interacting protein 5, receptor interacting protein kinase 5, Receptor-interacting serine/threonine-protein kinase 5, RIP5EC 2.7.12.1, RIPK5, sgK496, Sugen kinase 496 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?