missing translation for 'onlineSavingsMsg'
Learn More

RIPK3/RIP3 Antibody (1312A), DyLight 680, Novus Biologicals™

Product Code. 18417407 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417407 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417407 Supplier Novus Biologicals Supplier No. NBP276800FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RIPK3/RIP3 Monoclonal antibody specifically detects RIPK3/RIP3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPK3/RIP3
Applications Western Blot
Classification Monoclonal
Clone 1312A
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3
Host Species Rabbit
Immunogen RIPK3/RIP3 Antibody (1312A) [DyLight 680] was made to a partial synthetic peptide made to an internal portion of the human RIPK3/RIP3 protein (between amino acids 470-540). This seqeunce has 100% homology to isoform CRA_a and isoform CRA_b.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11035
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.