missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RIPK2, His Tag Recombinant Protein
RIPK2 is a serine/threonine kinase.
Brand: Invitrogen™ PV4213
Code : 50
Additional Details : Weight : 0.05250kg
Description
RIPK2 is a serine⁄threonine kinase that is involved in cell activation, proliferation, differentiation, and apoptosis.Specifications
NP_003812 | |
38.9 kDa | |
RIPK2, RICK, RIP2 | |
aa 1-299 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: TKL Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
receptor-interacting serine-threonine kinase 2 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction, Apoptosis |
Kinase Assay | |
10 μg | |
RIPK Kinase Family | |
Human | |
His Tag | |
Biochemical binding/kinetic | |
Dry Ice | |
RIPK2 | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |