missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK1/RIP1 Rabbit anti-Human, Clone: 2V4N8, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £396.00
Specifications
| Antigen | RIPK1/RIP1 |
|---|---|
| Clone | 2V4N8 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18315224
|
Novus Biologicals
NBP3-15786-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18343477
|
Novus Biologicals
NBP3-15786-100UL |
100 μg |
£396.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RIPK1/RIP1 Monoclonal antibody specifically detects RIPK1/RIP1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| RIPK1/RIP1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Necroptosis | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 8737 | |
| IgG | |
| Affinity purified |
| 2V4N8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 289-490 of human RIPK1/RIP1 (Q13546). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title