missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK1/RIP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£292.00 - £435.00
Specifications
| Antigen | RIPK1/RIP1 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18452191
|
Novus Biologicals
NBP1-87128-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18284667
|
Novus Biologicals
NBP1-87128 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RIPK1/RIP1 Polyclonal specifically detects RIPK1/RIP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RIPK1/RIP1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP | |
| RIPK1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 8737 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KEYSNENAVVKRMQSLQLDCVAVPSSRSNSATEQPGSLHSSQGLGMGPVEESWFAPSLEHPQEENEPSLQSKLQDEANYHLYGSRMDRQTKQQPRQNVAYNREEERRRRVSHDPFAQQRPYENFQNTEGKGTAYSSAASHGNAV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title