missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK1/RIP1 Antibody (CL11357), Novus Biologicals™
Mouse Monoclonal Antibody
£317.00 - £473.00
Specifications
| Antigen | RIPK1/RIP1 |
|---|---|
| Clone | CL11357 |
| Dilution | Western Blot 1 ug/ml |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613414
|
Novus Biologicals
NBP3-21190-25ul |
25 μg |
£317.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663704
|
Novus Biologicals
NBP3-21190-100ul |
100 μg |
£473.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RIPK1/RIP1 Monoclonal antibody specifically detects RIPK1/RIP1 in Human samples. It is validated for Western BlotSpecifications
| RIPK1/RIP1 | |
| Western Blot 1 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP | |
| This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of Q13546. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL11357 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Necroptosis | |
| PBS, pH 7.2, 40% glycerol | |
| 8737 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title