missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK1/RIP1 Antibody (CL11357) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21190MFV500
This item is not returnable.
View return policy
Description
RIPK1/RIP1 Monoclonal antibody specifically detects RIPK1/RIP1 in Human samples. It is validated for Western Blot
Specifications
| RIPK1/RIP1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP | |
| This antibody was developed using a recombinant protein derived from Q13546, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Necroptosis | |
| 8737 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Western Blot | |
| CL11357 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction