missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK1/RIP1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77077AF700
This item is not returnable.
View return policy
Description
RIPK1/RIP1 Polyclonal antibody specifically detects RIPK1/RIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
| RIPK1/RIP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Alexa Fluor 700 | |
| Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human RIPK1. The immunogen is located within amino acids 180 - 230 of RIPK1. Amino Acid Squence: DVNAKPTEKSDVYS | |
| 0.1 mL | |
| Apoptosis, Necroptosis | |
| 8737 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction