missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RIPA 56
GREENER_CHOICE

Product Code. 18708426
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18708426

Brand: Tocris Bioscience™ 7339/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and metabolically stable RIP1 kinase inhibitor

RIPA 56 is a potent and metabolically stable receptor-interacting protein kinase 1 (RIPK1) inhibitor (EC50 = 13 nM). RIPA-56 shows no inhibition of RIP3 kinase at 10 μM . RIPA 56 blocks necroptosis and aging of the mouse male reproductive system. RIPA 56 reduces tumor necrosis factor α (TNFα)-induced mortality and multi-organ damage in a mouse model of systemic inflammatory response syndrome (SIRS). RIPA 56 is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Hydroxy-2,2-dimethyl-N-(phenylmethyl)butanamide
CAS 1956370-21-0
Quantity 10 mg
Target RIP Kinase Inhibitors
Molecular Formula C13H19NO2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.