missing translation for 'onlineSavingsMsg'
Learn More

RIP5 Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-99509CL1

Product Code. 30158099

  • £622.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RIP5 Polyclonal antibody specifically detects RIP5 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RIP5
Polyclonal
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
55969
Store at 4°C in the dark. Do not freeze.
IgG
Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CoraFluor 1
C20orf24, chromosome 20 open reading frame 24, PNAS-11
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human RIP5.
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.