missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RIP3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA141533

Product Code. 11514530

  • £435.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence homology: Chimpanzee (100%). This antibody is not suitable for testing of mouse RIP3. Immunogen sequence: QEGPKDPEAW SRPQ.

RIPK3 is a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases, which contains a C-terminal domain unique from other RIP family members. RIPK3 is predominantly localized to the cytoplasm, and specifically, are colocalized in the mitochondrion which can undergo nucleocytoplasmic shuttling dependent on novel nuclear localization and export signals. RIPK3 is a component of the tumor necrosis factor (TNF) receptor-I signaling complex, and can induce apoptosis and weakly activate the NF-kappaB transcription factor and caspase-8 prevents RIPK3-dependent necrosis without inducing apoptosis by functioning in a proteolytically active complex with CFLAR and that this complex is required for the protective function.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RIP3
Polyclonal
Unconjugated
RIPK3
2610528K09Rik; AW107945; Hcyp2; Homocysteine respondent protein HCYP2; mRIP3; receptor interacting protein 3; receptor interacting serine/threonine kinase 3; receptor-interacting protein 3; receptor-interacting serine/threonine-protein kinase 3; receptor-interacting serine-threonine kinase 3; Rip3; RIP-3; Ripk3; RIP-like protein kinase 3
Rabbit
Antigen affinity chromatography
RUO
11035, 56532
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.05% sodium azide
Q9QZL0, Q9Y572
RIPK3
Synthetic peptide corresponding to amino acids 497-518 of human RIPK3/RIP3.
100 μg
Primary
Human, Mouse, Monkey
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.