missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIOK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103249
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RIOK3.
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. SUDD, also known as RIOK3 (RIO kinase 3), is a 519 amino acid protein that contains one protein kinase domain and belongs to the Ser/Thr protein kinase family. Expressed in a variety of tissues, SUDD catayzes the ATP-dependent phosphorylation of target proteins, thereby influencing signaling events throughout the cell. SUDD is expressed as two isoforms due to alternative splicing events.
Specifications
| RIOK3 | |
| Polyclonal | |
| Unconjugated | |
| RIOK3 | |
| 1200013N13Rik; D18Ertd331e; E130306C24Rik; homolog of the Aspergillus nidulans sudD gene product; RIO kinase 3; RIOK3; Serine/threonine-protein kinase RIO3; SUDD; sudD (suppressor of bimD6, Aspergillus nidulans) homolog; sudD homolog; sudD suppressor of Aspergillus nidulans bimD6 homolog; sudD suppressor of bimD6 homolog; sudD, suppressor of bimD6 homolog; testicular secretory protein Li 47 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 66878, 8780 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14730, Q9DBU3 | |
| RIOK3 | |
| A synthesized peptide derived from human RIOK3(Accession O14730), corresponding to amino acid residues E278-W328. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction