missing translation for 'onlineSavingsMsg'
Learn More

RIOK3 Antibody (3G11), Novus Biologicals™

Product Code. 18377449 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377449 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377449 Supplier Novus Biologicals Supplier No. H00008780M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RIOK3 Monoclonal antibody specifically detects RIOK3 in Human samples. It is validated for Western Blot, ELISA, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIOK3
Applications Western Blot, ELISA, Sandwich ELISA, KnockDown
Classification Monoclonal
Clone 3G11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH39729
Gene Alias DKFZp779L1370, EC 2.7.11.1, homolog of the Aspergillus nidulans sudD gene product, RIO kinase 3, RIO kinase 3 (yeast), serine/threonine-protein kinase RIO3, sudD (suppressor of bimD6, Aspergillus nidulans) homolog, sudD homolog, sudD suppressor of bimD6 homolog, SUDDsudD suppressor of Aspergillus nidulans bimD6 homolog
Host Species Mouse
Immunogen RIOK3 (AAH39729, 411 a.a. ~ 516 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NMLWHAGKVWLIDVSQSVEPTHPHGLEFLFRDCRNVSQKGGVKEALSERELFNAVSGLNITADNEADFLAEIEALEKMNEDHVQKNGRKAASFLKDDGDPPLLYDE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8780
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.