missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIOK2 Monoclonal Antibody (OTI5D11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526074
This item is not returnable.
View return policy
Description
RIOK2 Monoclonal Antibody for Western Blot, IHC (P), Flow
RIOK2 belongs to the protein kinase superfamily; RIO-type Ser/Thr kinase family. Serine/threonine protein kinases, such as RIOK2, phosphorylate the OH group of serine or threonine (which have similar sidechains). Activity of these protein kinases can be regulated by specific events (e.g. DNA damage), as well as numerous chemical signals, including cAMP/cGMP, Diacylglycerol, and Ca2+/calmodulin.
Specifications
| RIOK2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BVS4 | |
| RIOK2 | |
| Human recombinant protein fragment corresponding to amino acids 277-552 of RIOK2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5D11 | |
| Unconjugated | |
| RIOK2 | |
| 2010110K24Rik; 2410085M17Rik; RIO kinase 2; RIO kinase 2 (yeast); RIO2; Riok2; Serine/threonine-protein kinase RIO2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55781 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction