missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIOK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100684
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RIOK2.
RIOK2 belongs to the protein kinase superfamily; RIO-type Ser/Thr kinase family. Serine/threonine protein kinases, such as RIOK2, phosphorylate the OH group of serine or threonine (which have similar sidechains). Activity of these protein kinases can be regulated by specific events (e.g. DNA damage), as well as numerous chemical signals, including cAMP/cGMP, Diacylglycerol, and Ca2+/calmodulin.
Specifications
| RIOK2 | |
| Polyclonal | |
| Unconjugated | |
| RIOK2 | |
| 2010110K24Rik; 2410085M17Rik; RIO kinase 2; RIO kinase 2 (yeast); RIO2; Riok2; Serine/threonine-protein kinase RIO2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55781 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BVS4 | |
| RIOK2 | |
| A synthesized peptide derived from human RIOK2(Accession Q9BVS4), corresponding to amino acid residues E402-L452. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction