Learn More
RIOK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17222-1-AP-20UL
 View more versions of this product
Product Code. 16867074
  • £135.00 / 20µL
Description
FXR1 is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. It may regulate intracellular transport and local transltion of certain mRNAs. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predomitly with the 60S ribosomal subunit. FXR1 is required for embryonic and postnatal development of muscle tissue. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
RIOK1
Polyclonal
Unconjugated
RIOK1
AD034, RIO kinase 1, RIO kinase 1 (yeast), RIOK1
Rabbit
Antigen Affinity Chromatography
RUO
291061, 71340, 83732
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q922Q2, Q9BRS2
Riok1
RIOK1 Fusion Protein Ag11057
20 μL
Primary
Human, Rat, Mouse