missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIOK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562409
This item is not returnable.
View return policy
Description
Immunogen sequence: DWDWDEGVGK LAKGYVWNGG SNPQANRQTS DSSSAKMSTP ADKVLRKFEN KINLDKLNVT DSVINKVTEK SRQKEADMYR IKD Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
FXR1 is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. It may regulate intracellular transport and local transltion of certain mRNAs. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predominantly with the 60S ribosomal subunit. FXR1 is required for embryonic and postnatal development of muscle tissue. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| RIOK1 | |
| Polyclonal | |
| Unconjugated | |
| Riok1 | |
| 3110046C13Rik; 5430416A05Rik; AD034; bA288G3.1; RIO kinase 1; RIO kinase 1 (yeast); RIO1; Riok1; RRP10; Serine/threonine-protein kinase RIO1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83732 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BRS2 | |
| Riok1 | |
| Recombinant protein corresponding to Human RIOK1. Recombinant protein control fragment (Product #RP-99573). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction