Learn More
Elma™ Rinsing Sinks
Product Code. 17831307
Click to view available options
Model:
L
M
S
XL
Unit Size:
Each
Product Code. 17831307
Brand: Elma™ 1113839
Only null left
Add to basket
Add to basket
This rinsing sink removes any remaining contamination after cleaning.
  • Important addition to the overall cleaning process
  • Tabletop unit is available in four different sizes – the L and XL models feature an additional side drain
  • Once the cleaning process in the ultrasonic unit is complete, the basket can be inserted directly into the Elma Sink
  • Ensures that any residues and cleaning chemicals are rinsed off thoroughly
  • In conjunction with a suitable rinsing medium, this guarantees optimal cleaning results
Specifications
Type Sink
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Model S