Learn More
Ansell RINGERS Gloves 133
Product Code. 17858138 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
120 pairs
Product Code. 17858138
Brand: Ansell 13306
Only null left
Add to basket
Add to basket
RINGERS Gloves 133 are tactile and secure cuff automotive gloves.
  • Secure cuff with hook and loop TPR pull tab closure
  • Soft and durable premium mesh fabric on top of hand for optimal comfort and flexibility
  • Premium synthetic leather used for split palm construction for long lasting durability
  • Wrap around index finger protecting wear and tear zone
  • Lightweight fabric between fingers for excellent dexterity
  • Touchscreen compatible index, middle and thumb tips
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) ANSI ABR 3, ANSI Cut A1, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, Category II
Chlorinated No
Coating Material Uncoated
Color Black
Cut Resistance Rating ANSI A1
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size 6
Finger Style Full
For Use With (Application) General Purpose Auto Aftermarket Handling structural and body components Body panel repair and replacement Oil, fluids and filter change Engine transmission and repair Tire rotation and changes Equipment maintenance and repair Driving fork lifts and vehicles Delivery of parts to production line Changing tools and dies
Gender Unisex
Glove Cuff Style Velcro™