Learn More
Ansell Ringers 065 R-Flex Impact Glove
Product Code. 17657445 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
12
13
Unit Size:
12 pairs
Product Code. 17657445
Brand: Ansell 06507
Only null left
Add to basket
Add to basket
Ringers 065 R-Flex Impact Gloves are cut-resistant gloves, with a TPR design and knit shell for dexterous comfort.
  • The Ringers 065 R-Flex Impact glove with a Nitrile Half Dip palm combines protection against impact hazards with cut resistance for a wide range of applications.
  • A breathable knit shell developed with Ringers' F3 Technology™ and patented TPR design provide superior dexterity and comfort.
  • The sandy finish palm ensures secure grip in wet and dry environments, and touchscreen compatible index, middle and thumb tips allow technology interaction.
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Nitrile
Color Hi-vis Yellow, Gray
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material HPPE
Product Type Mechanical Protection Gloves
Size 7
Glove Cuff Style Knit