missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RING1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15037-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene belongs to the RING finger family, members of which encode proteins characterized by a RING domain, a zinc-binding motif related to the zinc finger domain. The gene product can bind DNA and can act as a transcriptional repressor. It is associated with the multimeric polycomb group protein complex. The gene product interacts with the polycomb group proteins BMI1, EDR1, and CBX4, and colocalizes with these proteins in large nuclear domains. It interacts with the CBX4 protein via its glycine-rich C-terminal domain. The gene maps to the HLA class II region, where it is contiguous with the RING finger genes FABGL and HKE4.Specifications
| RING1 | |
| Polyclonal | |
| Unconjugated | |
| RING1 | |
| Polycomb complex protein RING1, ring finger protein 1, RING1, RING1A, RNF1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19763, 309626, 6015 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35730, Q06587, Q6MGB6 | |
| RING1 | |
| RING1 Fusion Protein Ag6763 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction