missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RING1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582361
This item is not returnable.
View return policy
Description
Immunogen sequence: TRYVKTTGNA TVDHLSKYLA LRIALERRQQ QEAGEPGGPG GGASDTGGPD GCGGEGGGAG GGDGPEEPAL PSLEGVSEKQ YTIYIAPGGG AFTTLNGSLT LELVNEKFWK VS Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 94%.
This gene belongs to the RING finger family, members of which encode proteins characterized by a RING domain, a zinc-binding motif related to the zinc finger domain. The gene product can bind DNA and can act as a transcriptional repressor. It is associated with the multimeric polycomb group protein complex. The gene product interacts with the polycomb group proteins BMI1, EDR1, and CBX4, and colocalizes with these proteins in large nuclear domains. It interacts with the CBX4 protein via its glycine-rich C-terminal domain. The gene maps to the HLA class II region, where it is contiguous with the RING finger genes FABGL and HKE4.
Specifications
| RING1 | |
| Polyclonal | |
| Unconjugated | |
| RING1 | |
| E3 ubiquitin-protein ligase RING1; Polycomb complex protein RING1; really interesting new gene 1 protein; RING finger protein 1; RING1; Ring1A; RING-type E3 ubiquitin transferase RING1; Rnf1; Transcription repressor Ring1A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6015 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q06587 | |
| RING1 | |
| Recombinant protein corresponding to Human RING1. Recombinant protein control fragment (Product #RP-89328). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Spot an opportunity for improvement?Share a Content Correction