missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RING1 Antibody (8E8A2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £416.00
Specifications
| Antigen | RING1 |
|---|---|
| Clone | 8E8A2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672766
|
Novus Biologicals
NBP2-37319-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18163828
|
Novus Biologicals
NBP2-37319 |
0.1 mL |
£416.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RING1 Monoclonal specifically detects RING1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| RING1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 6.3.2, EC 6.3.2.-, Polycomb complex protein RING1, Really interesting new gene 1 protein, ring finger protein 1RING1A, RNF1E3 ubiquitin-protein ligase RING1 | |
| RING1 | |
| IgG1 | |
| Affinity Purified |
| 8E8A2 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Mouse | |
| Zinc Finger | |
| PBS with 0.05% Sodium Azide | |
| 6015 | |
| Purified recombinant fragment of human Ring1 (AA: 79-263) expressed in E. Coli. | |
| Primary | |
| 42.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title