missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ring finger protein 138 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-88154
This item is not returnable.
View return policy
Description
Ring finger protein 138 Polyclonal specifically detects Ring finger protein 138 in Human samples. It is validated for Western Blot.
Specifications
| Ring finger protein 138 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| E3 ubiquitin-protein ligase RNF138, EC 6.3.2.-, hNARF, HSD-4, MGC8758, NARF, Nemo-like kinase-associated RING finger protein, ring finger protein 138NLK-associated RING finger protein, STRIN | |
| The immunogen is a synthetic peptide directed towards the C terminal region of human Ring finger protein 138 | |
| 100 μL | |
| Zinc Finger | |
| 51444 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction