missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ring finger protein 138 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93425-0.02ml
This item is not returnable.
View return policy
Description
Ring finger protein 138 Polyclonal antibody specifically detects Ring finger protein 138 in Human samples. It is validated for Western Blot
Specifications
| Ring finger protein 138 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| E3 ubiquitin-protein ligase RNF138, EC 6.3.2.-, hNARF, HSD-4, MGC8758, NARF, Nemo-like kinase-associated RING finger protein, ring finger protein 138NLK-associated RING finger protein, STRIN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-245 of mouse Ring finger protein 138 (Q9CQE0). SVTRRERACPERALDLENIMRRFSGSCRCCSKKIKFYRMRHHYKSCKKYQDEYGVSSVIPNFKISQDSVRSSNRSETSASDNTETYQEDTSSSGHPTFKCPLCQESNFTRQRLLDHCNSNHLFQIVPVTCPICVSLPWGDPSQITRNFVSHLNQRHQFDYGEFVNLQLDEETQYQTAVEESFQVNM | |
| 0.02 mL | |
| Zinc Finger | |
| 51444 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction