missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12709-1-AP-150UL
This item is not returnable.
View return policy
Description
RIN3 is a member of the RIN family of Ras interaction-interference proteins, which are binding partners to the RAB5 small GTPases (Kajiho et al., 2003 ).Specifications
| RIN3 | |
| Polyclonal | |
| Unconjugated | |
| RIN3 | |
| Ras and Rab interactor 3, RIN3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217835, 314397, 79890 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P59729, Q8TB24 | |
| Rin3 | |
| RIN3 Fusion Protein Ag3390 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction