missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIN3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total RIN3.
RIN3 is a member of the RIN family of Ras interaction-interference proteins, which are binding partners to the RAB5 small GTPases (Kajiho et al., 2003 ).
Specifications
Specifications
| Antigen | RIN3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RIN3 |
| Gene Accession No. | Q8TB24 |
| Gene Alias | 6430500K07; C86933; RAB5 interacting protein 3; ras and Rab interactor 3; Ras interaction/interference protein 3; Rin3 |
| Gene Symbols | RIN3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?