missing translation for 'onlineSavingsMsg'
Learn More

RIN2 Antibody (1E6), Novus Biologicals™

Product Code. 18321959 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321959 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321959 Supplier Novus Biologicals Supplier No. H00054453M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RIN2 Monoclonal antibody specifically detects RIN2 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIN2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1E6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_061866
Gene Alias RAB5 interacting protein 2, Ras and Rab interactor 2, RAS association (RalGDS/AF-6) domain containing protein JC265, Ras association domain family 4, Ras inhibitor JC265, Ras interaction/interference protein 2, RASSF4MACS
Host Species Mouse
Immunogen RIN2 (NP_061866, 786 a.a. ~ 894 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DFQNYLRVAFQEVNSGCTGKTLLVRPYITTEDVCQICAEKFKVGDPEEYSLFLFVDETWQQLAEDTYPQKIKAELHSRPQPHIFHFVYKRIKNDPYGIIFQNGEEDLTT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54453
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.