Learn More
Invitrogen™ RIN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110298
Description
Immunogen sequence: MESPGESGAG SPGAPSPSSF TTGHLAREKP AQDPLYDVPN ASGGQAGGPQ RPGRVVSLRE RLLLTRPVWL QLQANAAAAL HMLRTEPPGT FLVRKSNTRQ CQALCMRLPE ASGPSFVSSH YILESPGGVS LEGSELMFPD LVQLICAYCH TRDILLLPLQ LPRAIHHAAT HKELEAISHL GIEFWSSSLN IKAQRGPAGG PVLPQLKARS PQELDQGTGA ALCFFNPLFP GDLGPTKREK FKRSFKVRVS TETSSPLSPP AVPPPPVPVL PGAVPSQTER.
Rit and its neuron-specific homologue, Rin, define a recently discovered subfamily of Ras-related GTPases. Rit and Rin are membrane-associated in spite of the fact that they lack a CAAX box or similar C-terminal lipidation motif. Rit and Rin display 64% amino acid sequence identity and share a unique nine amino acid effector domain (DPTIEDAYK) that is 100% conserved between the murine and human proteins. Although the effector domain sequences of Rit and Rin are very similar to that of Ras, Rit and Rin have been shown to interact with the known Ras-binding proteins RalGDS, Rlf and AF-6, but not the Raf kinases, RIN1 or the p110 subunit of PI3 kinase. For this reason, it has been suggested that Rit and Rin may play important roles in the regulation of signaling pathways distinct from those controlled by Ras.
Specifications
| RIN1 | |
| Polyclonal | |
| Unconjugated | |
| RIN1 | |
| Ras and Rab interactor 1; ras inhibitor; ras inhibitor 1; ras inhibitor JC99; ras inhibitor RIN1; ras interaction/interference protein 1; RIBA; Rin1; RIT2; ROC2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 207119, 225870 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.7 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P97680, Q921Q7 | |
| RIN1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human RIN1 (NP_004283.2). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.