missing translation for 'onlineSavingsMsg'
Learn More

RIN1 Antibody, Novus Biologicals™

Product Code. 18329058 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329058 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329058 Supplier Novus Biologicals Supplier No. H00009610B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

RIN1 Polyclonal antibody specifically detects RIN1 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIN1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_004283
Gene Alias Ras and Rab interactor 1, ras inhibitor 1, Ras inhibitor JC99, ras inhibitor RIN1, Ras interaction/interference protein 1
Host Species Mouse
Immunogen RIN1 (NP_004283, 1 a.a. - 783 a.a.) full-length human protein. MESPGESGAGSPGAPSPSSFTTGHLAREKPAQDPLYDVPNASGGQAGGPQRPGRVVSLRERLLLTRPVWLQLQANAAAALHMLRTEPPGTFLVRKSNTRQCQALCMRLPEASGPSFVSSHYILESPGGVSLEGSELMFPDLVQLICAYCHTRDILLLPLQLPRAIHHAATHKELEAISHLGIEFWSSSLNIKAQRGPAGGPVLPQLKARSPQELDQGTGAALCFFNPLFPGDLGPTKREKFKRSFKVRVSTETSSPLSPPAVPPPPVPVLPGAVPSQTERLPPCQLLRRESSVGYRVPAGSGPSLPPMPSLQEVDCGSPSSSEEEGVPGSRGSPATSPHLGRRRPLLRSMSAAFCSLLAPERQVGRAAAALMQDRHTAAGQLVQDLLTQVRAGPEPQELQGIRQALSRARAMLSAELGPEKLLSPKRLEHVLEKSLHCSVLKPLRPILAARLRRRLAADGSLGRLAEGLRLARAQGPGAFGSHLSLPSPVELEQVRQKLLQLLRTYSPSAQVKRLLQACKLLYMALRTQEGEGAGADEFLPLLSLVLAHCDLPELLLEAEYMSELLEPSLLTGEGGYYLTSLSASLALLSGLGQAHTLPLSPVQELRRSLSLWEQRRLPATHCFQHLLRVAYQDPSSGCTSKTLAVPPEASIATLNQLCATKFRVTQPNTFGLFLYKEQGYHRLPPGALAHRLPTTGYLVYRRAEWPETQGAVTEEEGSGQSEARSRGEEQGCQGDGDAGVKASPRDIREQSETTAEGGQGQAQEGPAQPGEPEAEGSRAAEE
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9610
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.