missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIN1 Antibody (27G2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-66642
This item is not returnable.
View return policy
Description
RIN1 Monoclonal specifically detects RIN1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| RIN1 | |
| Monoclonal | |
| Unconjugated | |
| 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol (pH 7.5) with 3mM Sodium Azide | |
| Ras and Rab interactor 1, ras inhibitor 1, Ras inhibitor JC99, ras inhibitor RIN1, Ras interaction/interference protein 1 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| Signal Transduction | |
| 9610 | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 27G2 | |
| Western Blot | |
| Q13671 | |
| RIN1 | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with full-length recombinant human Rin protein and mouse myeloma cells. | |
| 100 μg | |
| Primary | |
| Does not cross-react with Rit protein. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction