missing translation for 'onlineSavingsMsg'
Learn More

RIMS4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18342161 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342161 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342161 Supplier Novus Biologicals Supplier No. NBP2834470.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RIMS4 Polyclonal specifically detects RIMS4 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIMS4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias C20orf190, chromosome 20 open reading frame 190, dJ781B1.3, Rab3-interacting molecule 4, regulating synaptic membrane exocytosis 4, regulating synaptic membrane exocytosis protein 4, RIM 4, RIM4, RIM4 gamma, RIM4gamma
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of human RIMS4
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140730
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.