missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIMS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599707
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Rim4.
Regulates synaptic membrane exocytosis. Also a member of a family of synaptic proteins involved in neurotransmitter release.
Specifications
| RIMS4 | |
| Polyclonal | |
| Unconjugated | |
| Rims4 | |
| 6530406L02; C20orf190; rab3-interacting molecule 4; regulating synaptic membrane exocytosis 4; regulating synaptic membrane exocytosis protein 4; RIM 4; RIM4; RIM-4; RIM4 gamma; RIM4gamma; RIM4-gamma; RIMS4; synaptic regulatory protein RIM4gamma | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140730, 266976 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8CIX1, Q9H426 | |
| Rims4 | |
| A synthesized peptide derived from human RIMS4(Accession Q9H426), corresponding to amino acid residues A32-Y82. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction