missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIMS2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-41077CL1
This item is not returnable.
View return policy
Description
RIMS2 Polyclonal antibody specifically detects RIMS2 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| RIMS2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| DKFZp781A0653, KIAA0751RIM2OBOERAB3 interacting protein 3, non-small cell lung cancer RimL3a protein, non-small cell lung cancer RimL3c protein, nuclear protein, rab3-interacting molecule 2, Rab-3-interacting molecule 2, Rab-3-interacting protein 3, RAB3IP3, regulating synaptic membrane exocytosis 2, regulating synaptic membrane exocytosis protein 2, RIM 2 | |
| Antibody was raised against a 16 amino acid peptide near the carboxy terminusof human RIM2. The immunogen is located within amino acids 30 - 80 of RIM2. Amino Acid Squence: MDRQKKEEEKEQSVLK | |
| 0.1 mL | |
| Neuroscience | |
| 9699 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction