missing translation for 'onlineSavingsMsg'
Learn More

Rimantadine Hydrochloride, TRC

Product Code. 30138092
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138092 1 g 1g
17901765 10 mg 10mg
17982185 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138092 Supplier Toronto Research Chemicals Supplier No. TRCR5170001G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1501-84-4
Molecular Formula C12 H21 N . Cl H
Molecular Weight (g/mol) 215.76
Synonym Tricyclo[3.3.1.13,7]decane-1-methanamine, α-methyl-, hydrochloride (1:1), 1-Adamantanemethylamine, α-methyl-, hydrochloride (7CI,8CI), Tricyclo[3.3.1.13,7]decane-1-methanamine, α-methyl-, hydrochloride (9CI), 1-(1-Adamantyl)ethylamine hydrochloride, 1-(1-Aminoethyl)adamantane hydrochloride, EXP 126, Flumadine, JP 61, Meradan, Meradane, NSC 206764, Remantadin, Remantadine, Rimantadine hydrochloride, Roflual, α-Methyl-1-adamantanemethylamine hydrochloride
IUPAC Name 1-(1-adamantyl)ethanamine;hydrochloride
SMILES Cl.CC(N)C12CC3CC(CC(C3)C1)C2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H21N.ClH/c1-8(13)12-5-9-2-10(6-12)4-11(3-9)7-12;/h8-11H,2-7,13H2,1H3;1H
Formula Weight 215.1441
Percent Purity >95%
Chemical Name or Material Rimantadine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.