missing translation for 'onlineSavingsMsg'
Learn More

Rimacalib, MedChemExpress

Product Code. 16482646
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482646 25 mg 25mg
16455897 5 mg 5mg
16439716 50 mg 50mg
16423168 10 mg 10mg
16404456 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16482646 Supplier Medchem Express Supplier No. HY100779
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rimacalib (SMP 114) is a Ca2+/calmodulin-dependent protein kinase II (CaMKII) inhibitor, with IC50s of ∼1 μM for CaMKIIα to ∼30 μM for CaMKIIγ.

Rimacalib (SMP 114) is a Ca2+/calmodulin-dependent protein kinase II (CaMKII) inhibitor, with IC50s of ∼1 μM for CaMKIIα to ∼30 μM for CaMKIIγ.

Specifications

Chemical Name or Material Rimacalib
CAS 215174-50-8
Color Pink
Purity Grade Notes Research
Molecular Formula C22H23FN4O2
Quantity 25 mg
Synonym SMP 114
Solubility Information DMSO : 125 mg/mL (316.90 mM; Need ultrasonic)
SMILES N=C(N1CCOCC1)NC2=CC([C@H](C3=CC=C(C4=CC=CC=C4)C(F)=C3)C)=NO2
Molecular Weight (g/mol) 394.44
Formula Weight 394.44
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.