missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RIM3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12858652
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12858652 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12858652 Supplier Invitrogen™ Supplier No. PA520475

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-20475 can be used with blocking peptide PEP-0595.

Rab3-interacting molecules (RIMs) are synaptic proteins are synaptic proteins necessary for neural transmission and plasticity. While both Rim1 and Rim 2 are thought to be effector proteins for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner, less is known of Rim3. Expression of Rim3 in PC12 cells induced a significant increase in calcium-triggered exocytosis, with no appreciable change in the baseline release, suggesting that it plays a role in the regulation of exocytosis. Rim3 protein localizes primarily to neuronal dendrites and the postsynaptic densities, as opposed to Rim1 which is found in presynapse locations, indicating that Rim3 may contribute to synapse transmission and plasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIM3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene RIMS3
Gene Accession No. Q80U57, Q9UJD0
Gene Alias A730060M23Rik; KIAA0237; mKIAA0237; Nim2; NIM3; Rab-3 interacting molecule 3; rab-3-interacting molecule 3; regulating synaptic membrane exocytosis 3; regulating synaptic membrane exocytosis protein 3; RIM 3; RIM3; RIM3 gamma; Rims3; RP4-739H11.2; si:ch211-239d6.1
Gene Symbols RIMS3
Host Species Rabbit
Immunogen A 17 amino acid peptide from near the amino terminus of human Rim3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242662, 9783
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.