missing translation for 'onlineSavingsMsg'
Learn More

Rilzabrutinib, MedChemExpress

Product Code. 16474588
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474588 10 mg 10mg
16465436 100 mg 100mg
16460887 50 mg 50mg
16433146 25 mg 25mg
16427307 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16474588 Supplier Medchem Express Supplier No. HY112166
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Rilzabrutinib (PRN1008) is a reversible covalent, selective and oral active inhibitor of Bruton’s Tyrosine Kinase (BTK), with an IC50 of 1.3 nM.

Rilzabrutinib (PRN1008) is a reversible covalent, selective and oral active inhibitor of Bruton’s Tyrosine Kinase (BTK), with an IC50 of 1.3 nM.

Specifications

Chemical Name or Material Rilzabrutinib
CAS 1575596-29-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C36H40FN9O3
Quantity 10 mg
Synonym PRN1008
Solubility Information DMSO : ≥ 130 mg/mL (195.27 mM)
SMILES NC1=C2C(C3=C(C=C(OC4=CC=CC=C4)C=C3)F)=NN([C@H]5CN(CCC5)C(/C(C#N)=C/C(C)(N6CCN(C7COC7)CC6)C)=O)C2=NC=N1
Molecular Weight (g/mol) 665.76
Formula Weight 665.76
Percent Purity 95.4%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.