missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RILPL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534359
This item is not returnable.
View return policy
Description
A suggested positive control is A549 cell lysate. PA5-34359 can be used with blocking peptide PEP-1402.
Does not regulate lysosomal morphology and distribution.
Specifications
| RILPL2 | |
| Polyclonal | |
| Unconjugated | |
| Rilpl2 | |
| p40phox-binding protein; Rab interacting lysosomal protein like 2; Rab interacting lysosomal protein-like 2; rab-interacting lysosomal protein-like 2; Rab-interacting lysosomal-like protein 2; Rilpl2; RILP-like protein 2; RLP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 196383 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q969X0 | |
| Rilpl2 | |
| An 16 amino acid synthetic peptide near the center of human RLP2. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction