missing translation for 'onlineSavingsMsg'
Learn More

RILPL2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18333780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333780 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333780 Supplier Novus Biologicals Supplier No. NBP281993

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RILPL2 Polyclonal specifically detects RILPL2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RILPL2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 2.5 ug/mL, Immunocytochemistry/Immunofluorescence 2.5 ug/ml, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ30380, FLJ32372, MGC7036, p40phox-binding protein, Rab interacting lysosomal protein-like 2, Rab-interacting lysosomal protein-like 2, RILP-like protein 2, RLP2
Gene Symbols RILPL2
Host Species Rabbit
Immunogen RILPL2 antibody was raised against an 16 amino acid synthetic peptide near the center of human RILPL2. The immunogen is located within amino acids 110 - 160 of RILPL2. Amino Acid Squence: VDLTDPNRPRFTLQEL
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196383
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.