missing translation for 'onlineSavingsMsg'
Learn More

Rilpivirine, TRC

Product Code. 30011211
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011211 2.5 mg 2.5mg
30005465 25 mg 25mg
30010783 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011211 Supplier Toronto Research Chemicals Supplier No. TRCR5098002.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500287-72-9
Molecular Formula C22 H18 N6
Molecular Weight (g/mol) 366.42
Synonym 4-[[4-[[4-[(1E)-2-Cyanoethenyl]-2,6-dimethylphenyl]amino]-2-pyrimidinyl]amino]benzonitrile, Edurant, R 278474, Rilpivirine, TMC 278
IUPAC Name 4-[[4-[4-[(E)-2-cyanoethenyl]-2,6-dimethylanilino]pyrimidin-2-yl]amino]benzonitrile
SMILES Cc1cc(\C=C\C#N)cc(C)c1Nc2ccnc(Nc3ccc(cc3)C#N)n2

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C22H18N6/c1-15-12-18(4-3-10-23)13-16(2)21(15)27-20-9-11-25-22(28-20)26-19-7-5-17(14-24)6-8-19/h3-9,11-13H,1-2H3,(H2,25,26,27,28)/b4-3+
Formula Weight 366.1593
Percent Purity >95%
Chemical Name or Material Rilpivirine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.