missing translation for 'onlineSavingsMsg'
Learn More

Rilpivirine, MedChemExpress

Product Code. 16490407
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490407 50 mg 50mg
16434058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490407 Supplier Medchem Express Supplier No. HY10574

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rilpivirine (R278474) is a potent and specific diarylpyrimidine (DAPY) non-nucleoside reverse transcriptase inhibitor (NNRTI). Rilpivirine has high antiviral activity against wild-type HIV (EC50=0.4 nM) and mutant viruses (EC50=0.1-2.0 nM). Rilpivirine has a high genetic barrier to resistance development of HIV.

Rilpivirine (R278474) is a potent and specific diarylpyrimidine (DAPY) non-nucleoside reverse transcriptase inhibitor (NNRTI). Rilpivirine has high antiviral activity against wild-type HIV (EC50=0.4 nM) and mutant viruses (EC50=0.1-2.0 nM). Rilpivirine has a high genetic barrier to resistance development of HIV.

Specifications

Chemical Name or Material Rilpivirine
Physical Form Solid
CAS 500287-72-9
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H18N6
For Use With (Application) COVID-19-anti-virus
Synonym R278474 TMC278 DB08864
Solubility Information DMSO : 50 mg/mL (136.46 mM; Need ultrasonic)
SMILES CC1=CC(/C=C/C#N)=CC(C)=C1NC2=CC=NC(NC3=CC=C(C=C3)C#N)=N2
Molecular Weight (g/mol) 366.42
Formula Weight 366.42
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.