missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Rilpivirine

Description
Rilpivirine is a potent diarylpyrimidine (DAPY) non-nucleoside reverse transcriptase inhibitor (NNRTI). It has high antiviral activity against wild-type HIV-1 and mutant viruses (EC50 values are 0.4 and 0.1-2.0 nM, respectively in a cell-based assay). This compound is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 4-[[4-[[4-[(1 E)-2-Cyanoethenyl]-2,6-dimethylphenyl]amino]-2-pyrimidinyl]amino]benzonitrile |
| CAS | 500287-72-9 |
| Quantity | 10 mg |
| Target | RNA/DNA Polymerase Inhibitors |
| Molecular Formula | C22H18N6 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?