missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RILP Polyclonal antibody specifically detects RILP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | RILP |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | rab-interacting lysosomal protein, PP10141, Rab interacting lysosomal protein |
| Host Species | Rabbit |
| Immunogen | RILP antibody was raised against an 18 amino acid synthetic peptide near the center of human RILP. The immunogen is located within amino acids 110 - 160 of RILP. Amino Acid Squence: DELRAHNRDLRQRGQETE |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?