missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RILP Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81991G
This item is not returnable.
View return policy
Description
RILP Polyclonal antibody specifically detects RILP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| RILP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 83547 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 488 | |
| rab-interacting lysosomal protein, PP10141, Rab interacting lysosomal protein | |
| RILP antibody was raised against an 18 amino acid synthetic peptide near the center of human RILP. The immunogen is located within amino acids 110 - 160 of RILP. Amino Acid Squence: DELRAHNRDLRQRGQETE | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction