missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Rilmenidine hemifumarate
GREENER_CHOICE

Product Code. 18747316
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18747316 25 mg 25mg
18718249 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18747316 Supplier Tocris Bioscience™ Supplier No. 0790/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

α2 agonist. Also I1 ligand

Rilmenidine hemifumarate is an I1-imidazoline binding site selective ligand and α2-adrenoceptor agonist. Possesses greater I1 vs α2 selectivity than the prototypical compound, clonidine. Also thought to enhance autophagy; shown to increase LC3-II levels in PC12 cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[N-(Dicyclopropylmethyl)amino]oxazoline hemifumarate
CAS 207572-68-7
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C10H16N2O.½C4H4O4
Formula Weight Observed MW: 238.29
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.